
Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate 95%
Catalog # APOSPC9397-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.66 g/mol
Melting Pt:53…55 °C
:GHS07
Storage Temperature:Ambient
CAS Number:4415-09-2
Formula:C₁₂H₉ClFNO₃
MDL Number:MFCD00214743