Order Entry
Italy
ContactUsLinkComponent
2-{[(4-Fluorophenyl)sulphonyl]amino}-2-phenylacetic acid 97%
2-{[(4-Fluorophenyl)sulphonyl]amino}-2-phenylacetic acid 97%
Catalog # APOSPC9374-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-{[(4-Fluorophenyl)sulphonyl]amino}-2-phenylacetic acid 97%
Catalog # APOSPC9374-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :309.32 g/mol
Melting Pt:179…181 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:117309-49-6
Formula:C₁₄H₁₂FNO₄S
MDL Number:MFCD00173006