2-(4-Fluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338982-40-4
:
Formula:C₁₀H₅FN₄O₂
MDL Number:MFCD00794272
:232.18 g/mol
Melting Pt:242…244 °C
Storage Temperature:Ambient
Frequently Bought Together