Order Entry
Italy
ContactUsLinkComponent
2-(Difluoromethoxy)phenylacetic acid 98%
2-(Difluoromethoxy)phenylacetic acid 98%
Catalog # APOSPC9251-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Difluoromethoxy)phenylacetic acid 98%
Catalog # APOSPC9251-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:86867-68-7
Formula:C₉H₈F₂O₃
MDL Number:MFCD00236237