2-Chloro-4-(trifluoromethyl)nicotinic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:590371-81-6
:
Formula:C₇H₃ClF₃NO₂
MDL Number:MFCD08458125
Melting Pt:157…160 °C
:225.56 g/mol
Storage Temperature:Ambient
Frequently Bought Together