Order Entry
Italy
ContactUsLinkComponent
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate 98%
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate 98%
Catalog # APOSPC906432-1G
Supplier:  Apollo Scientific
CAS Number:  
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate 98%
Catalog # APOSPC906432-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :672.12 g/mol
Storage Temperature:Ambient
CAS Number:77758-84-0
Formula:C₁₃H₅F₁₆IO₃S
MDL Number:MFCD00135119