Order Entry
Italy
ContactUsLinkComponent
4-(5-Fluoropyridin-2-yl)morpholine 98%
4-(5-Fluoropyridin-2-yl)morpholine 98%
Catalog # APOSPC900590-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(5-Fluoropyridin-2-yl)morpholine 98%
Catalog # APOSPC900590-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.2 g/mol
Storage Temperature:Ambient
CAS Number:1287217-51-9
Formula:C₉H₁₁FN₂O
MDL Number:MFCD18089238