
3-Bromo-2-fluorobenzoic acid 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:161957-56-8
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD00665763
:219.01 g/mol
Melting Pt:165…170 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together