Order Entry
Italy
ContactUsLinkComponent
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride 97%
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride 97%
Catalog # APOSPC8687-100G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2-(trifluoroacetyl)aniline hydrochloride 97%
Catalog # APOSPC8687-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :260.04 g/mol
Storage Temperature:Ambient
CAS Number:173676-59-0
 :MFCD09841379
Formula:C₈H₆Cl₂F₃NO
MDL Number:MFCD09263480