Order Entry
Italy
ContactUsLinkComponent
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Catalog # APOSPC8683-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(3-Methoxyphenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Catalog # APOSPC8683-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :337.26 g/mol
Storage Temperature:Ambient
CAS Number:524036-15-5
Formula:C₁₅H₁₀F₃N₃O₃
MDL Number:MFCD02253965