Ethyl-2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:955975-47-0
:
Formula:C₁₁H₁₀F₃N₃O₂S
MDL Number:MFCD03305618
:305.28 g/mol
Melting Pt:109…110 °C
Storage Temperature:Ambient
Frequently Bought Together