Order Entry
Italy
ContactUsLinkComponent
5-(Chloromethyl)-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole
Irritant
5-(Chloromethyl)-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole
Catalog # APOSPC8583-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(Chloromethyl)-2-[3-fluoro-4-(trifluoromethyl)phenyl]-4-methyl-1,3-thiazole
Catalog # APOSPC8583-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :309.71 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:317319-33-8
Formula:C₁₂H₈ClF₄NS
MDL Number:MFCD07369415