0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
4-Fluoro-N-methylbenzenesulphonamide
4-Fluoro-N-methylbenzenesulphonamide
4-Fluoro-N-methylbenzenesulphonamide
APOSPC8577-1G
:
Apollo Scientific
:
4-Fluoro-N-methylbenzenesulphonamide
APOSPC8577-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
56…58 °C
:
189.2073 g/mol
:
GHS07
:
Ambient
:
433-14-7
:
C₇H₈FNO₂S
:
MFCD01211989