Order Entry
Italy
ContactUsLinkComponent
2-Bromo-3'-fluoro-4'-(trifluoromethyl)acetophenone
Corrosive
2-Bromo-3'-fluoro-4'-(trifluoromethyl)acetophenone
Catalog # APOSPC8501-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Bromo-3'-fluoro-4'-(trifluoromethyl)acetophenone
Catalog # APOSPC8501-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:29…31 °C
Storage Temperature:Refrigerator
UN:1759
ADR:8,III
 :GHS05
CAS Number:544429-22-3
Formula:C₉H₅BrF₄O
MDL Number:MFCD03094511