Order Entry
Italy
ContactUsLinkComponent
3-(4-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
3-(4-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Catalog # APOSPC8496-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Catalog # APOSPC8496-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.3 g/mol
Melting Pt:>250 °C
Storage Temperature:Ambient
CAS Number:1547-15-5
Formula:C₁₄H₉FN₂OS
MDL Number:MFCD00807054
Melting Pt:>250 °C