Ethyl-4-chloro-1-(4-fluorophenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339031-90-2
:
Formula:C₁₃H₁₀ClFN₂O₃
MDL Number:MFCD00243751
Melting Pt:126…128 °C
:296.69 g/mol
Storage Temperature:Ambient
Frequently Bought Together