
2-Cyano-N-[3-(trifluoromethyl)phenyl]acetamide 97%
Catalog # APOSPC8448-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:143…145 °C
:228.18 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1960-77-6
Formula:C₁₀H₇F₃N₂O
MDL Number:MFCD00117941