
1,2-Bis[2'-(trifluoromethyl)phenyl]ethane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:287172-66-1
:
Formula:C₁₆H₁₂F₆
MDL Number:MFCD01631639
Melting Pt:74…76 °C
:318.26 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together