
2-Bromo-2',4'-bis(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:435271-21-9
:
Formula:C₁₀H₅BrF₆O
MDL Number:MFCD03094502
Melting Pt:50…52 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together