
4,5-Dichloro-2-fluorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:289039-49-2
:
Formula:C₇H₃Cl₂FO₂
MDL Number:MFCD00672988
:209.0019 g/mol
Melting Pt:148…150 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together