Order Entry
Italy
ContactUsLinkComponent
 
Irritant
2,6-Dibromo-4-fluorophenol 97%
Catalog # APOSPC8177-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dibromo-4-fluorophenol 97%
Catalog # APOSPC8177-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :269.9 g/mol
Melting Pt:54…56 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:344-20-7
Formula:C₆H₃Br₂FO
MDL Number:MFCD00002317