
6,7-Dichloro-3-(trifluoromethyl)-2-quinoxalinecarboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:951884-93-8
:
Formula:C₁₀H₃Cl₂F₃N₂O₂
MDL Number:MFCD09800927
:GHS07
Storage Temperature:Ambient
Frequently Bought Together