
5-(Trifluoromethyl)-1H-indazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885271-64-7
:
Formula:C₈H₅F₃N₂
MDL Number:MFCD08234904
:186.1300 g/mol
Melting Pt:123…125 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together