
4-(Trifluoromethyl)benzoic acid hydrazide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339-59-3
:
Formula:C₈H₇F₃N₂O
MDL Number:MFCD00051703
:204.15 g/mol
Melting Pt:115…121 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together