Order Entry
Italy
ContactUsLinkComponent
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one 98%
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one 98%
Catalog # APOSPC7913-1G
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one 98%
Catalog # APOSPC7913-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.300 g/mol
Storage Temperature:Ambient
CAS Number:477857-25-3
Formula:C₉H₃Cl₂F₃N₂O
MDL Number:MFCD09801051