
4-Hydroxy-3-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220239-68-9
:
Formula:C₈H₅F₃O₃
MDL Number:MFCD01091008
:206.12 g/mol
Melting Pt:144…147 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together