Order Entry
Italy
ContactUsLinkComponent
Ethyl-2-(pyrrolidin-1-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Ethyl-2-(pyrrolidin-1-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Catalog # APOSPC7558-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2-(pyrrolidin-1-yl)-4-trifluoromethyl-1,3-thiazole-5-carboxylate
Catalog # APOSPC7558-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.3 g/mol
Storage Temperature:Ambient
CAS Number:1000339-79-6
Formula:C₁₁H₁₃F₃N₂O₂S
MDL Number:MFCD09750913