
5-(Trifluoromethyl)anthranilic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:83265-53-6
:
Formula:C₈H₆F₃NO₂
MDL Number:MFCD01569537
:205.14 g/mol
Melting Pt:177 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together