2-Bromo-4-trifluoromethyl-1,3-thiazole-5-carboxylic acid
Catalog # APOSPC7487-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.04 g/mol
Melting Pt:130…132 °C
Storage Temperature:Ambient
CAS Number:162651-07-2
Formula:C₅HBrF₃NO₂S
MDL Number:MFCD09264552