Order Entry
Italy
ContactUsLinkComponent
4'-Trifluoromethoxyacetophenone 98%
Irritant
4'-Trifluoromethoxyacetophenone 98%
Catalog # APOSPC7433E-1KG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-Trifluoromethoxyacetophenone 98%
Catalog # APOSPC7433E-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :204.15
Boiling Pt:47…49 °C/15mmHg
Density:1.278 g/cm³ (25 °C)
Flash Pt:87 °C
EINECS:285-066-0
 :GHS07
Storage Temperature:Ambient
CAS Number:85013-98-5
Formula:C₉H₇F₃O₂
MDL Number:MFCD00042404