
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbaldehyde
Catalog # APOSPC7216-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:301.65 g/mol
Boiling Pt:106 °C/0.075mmHg
:GHS07
Storage Temperature:Ambient
CAS Number:950994-19-1
Formula:C₁₃H₇ClF₃NO₂
MDL Number:MFCD09607890