4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]benzenecarbaldehyde
Catalog # APOSPC7215-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:285.66 g/mol
Melting Pt:71…73 °C
Storage Temperature:Ambient
CAS Number:952182-72-8
Formula:C₁₃H₇ClF₃NO
MDL Number:MFCD09607889