Tetramethylammonium hexafluorophosphate 99%
Catalog # APOSPC6937-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:219.11 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
CAS Number:558-32-7
Formula:C₄H₁₂F₆NP
MDL Number:MFCD00012012
Melting Pt:>300 °C