Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2-(difluoromethyl)-1-fluorobenzene
4-Bromo-2-(difluoromethyl)-1-fluorobenzene 97%
4-Bromo-2-(difluoromethyl)-1-fluorobenzene 97%
4-Bromo-2-(difluoromethyl)-1-fluorobenzene 97%
APOSPC6822-25G
:
Apollo Scientific
:
4-Bromo-2-(difluoromethyl)-1-fluorobenzene 97%
APOSPC6822-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
225.01 g/mol
:
Ambient
:
445303-69-5
:
C₇H₄BrF₃
:
MFCD06411633