Order Entry
Italy
ContactUsLinkComponent
1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether 98%
Irritant
1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether 98%
Catalog # APOSPC6787-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether 98%
Catalog # APOSPC6787-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :253.38 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:32778-09-9
Formula:C₃Cl₃F₅O
MDL Number:MFCD01320777