Tetrabutylammonium trifluoromethanesulphonate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:35895-70-6
:
Formula:C₁₇H₃₆F₃NO₃S
MDL Number:MFCD00042585
:391.54 g/mol
Melting Pt:112…113 °C
Storage Temperature:Ambient
Frequently Bought Together