5-(3-Chlorophenyl)-2-(trifluoromethyl)-3-furoic acid
Catalog # APOSPC6439-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:290.63 g/mol
Melting Pt:164…166 °C
Storage Temperature:Ambient
CAS Number:242812-02-8
Formula:C₁₂H₆ClF₃O₃
MDL Number:MFCD01313475