
(S)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24435-45-8
:
Formula:C₄H₅F₃O₃
MDL Number:MFCD03092898
:158.08 g/mol
Boiling Pt:189 °C
Melting Pt:100 °C
Flash Pt:103 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together