

(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine
Catalog # APOSPC6382-50G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:224.28 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
:GHS07
CAS Number:112914-13-3
Formula:C₁₂H₁₇FN₂O
MDL Number:MFCD06658145