
1-Methyl-5-(trifluoromethyl)-1H-indazol-3-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5685-69-8
:
Formula:C₉H₈F₃N₃
MDL Number:MFCD07777192
Melting Pt:140…142 °C
UN:2811
ADR:6.1,III
:215.18 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together