
Praseodymium trifluoride, anhydrous 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13709-46-1
:
Formula:F₃Pr
MDL Number:MFCD00016287
Melting Pt:1395 °C
Density:6.2 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:237-254-9
UN:3288
ADR:6.1,III
:197.9 g/mol
:GHS07
Frequently Bought Together