Order Entry
Italy
ContactUsLinkComponent
2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) 97%
Irritant
2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) 97%
Catalog # APOSPC6285-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) 97%
Catalog # APOSPC6285-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Boiling Pt:90 °C/1.5mmHg
Density:1.506 g/cm³ (25 °C)
 :314.18 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:791-22-0
Formula:C₁₀H₁₀F₈O₂
MDL Number:MFCD01320774