
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine
Catalog # APOSPC6045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:283.30 g/mol
Melting Pt:75…80 °C
:GHS07
Storage Temperature:Ambient
CAS Number:114873-01-7
Formula:C₁₄H₁₈FNO₄
MDL Number:MFCD00672522