(R)-3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (R)-2-Hydroxy-2-(trifluoromethyl)propionic Acid, (R)-3,3,3-Trifluoro-2-hydroxyisobutyric Acid, (R)-3,3,3-Trifluoro-2-hydroxy-2-methylpropanoic acid
CAS Number:44864-47-3
:
Formula:C₄H₅F₃O₃
MDL Number:MFCD03095400
Boiling Pt:189 °C
Molecular Weight:158.08 g/mol
Melting Pt:108…110 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8 III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together