

Decafluoro-5,6-bis(trifluoromethyl)cyclohexane 70%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306-98-9
:
Formula:C₈F₁₆
MDL Number:MFCD00066612
:400.06 g/mol
Boiling Pt:101…103 °C
Density:1.861 g/cm³ (25 °C)
Flash Pt:23.9 °C
Storage Temperature:Ambient
EINECS:206-195-0
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together