
4-Bromo-3-(trifluoromethyl)benzaldehyde 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:34328-47-7
:
Formula:C₈H₄BrF₃O
MDL Number:MFCD08059130
:253.0200 g/mol
Melting Pt:52…54 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together