Order Entry
Italy
Orders LinkContactUsLinkComponent
1-(Pentafluorophenyl)ethanol 97%

1-(Pentafluorophenyl)ethanol 97%

Bottle, 1 g
Catalog # APOSPC5670-1G
Irritant
Supplier:   Apollo Scientific
1-(Pentafluorophenyl)ethanol 97%
Catalog # APOSPC5670-1G
Supplier:  Apollo Scientific
CAS Number:  830-50-2
1-(Pentafluorophenyl)ethanol 97%

Bottle, 1 g

CAS Number:  830-50-2
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (1-Hydroxyethyl)pentafluorobenzene
  • CAS Number:830-50-2
  • Formula:C₈H₅F₅O
  • MDL Number:MFCD00004510
  • Molecular Weight:212.12 g/mol
  • Boiling Pt:68…70 °C/30mmHg
  • Melting Pt:32…34 °C
  • Flash Pt:87 °C
  • Storage Temperature:Ambient
  • EINECS:212-597-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.