

Methyl-2,5-difluorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:362601-90-9
:
Formula:C₈H₆F₂O₂
MDL Number:MFCD03093800
:172.13 g/mol
Boiling Pt:202…204 °C
Density:1.265 g/cm³ (25 °C)
Flash Pt:80 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together