Order Entry
Italy
Orders LinkContactUsLinkComponent
2,3'-Dibromo-4'-fluoroacetophenone

2,3'-Dibromo-4'-fluoroacetophenone

Irritant
Supplier:   Apollo Scientific
2,3'-Dibromo-4'-fluoroacetophenone
Supplier:  Apollo Scientific
Certificates
2,3'-Dibromo-4'-fluoroacetophenone

3 Options Available Below

Product Details & Documents
Synonyms:  2-Bromo-1-(3-bromo-4-fluorophenyl)ethanone
  • CAS Number:435273-49-7
  • Formula:C₈H₅Br₂FO
  • MDL Number:MFCD07368759
  • Molecular Weight:295.93 g/mol
  • Melting Pt:48…52 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • UN:3335
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 23,20 €
    23,20 €
    1 * 1 g
       23,20 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 65,20 €
    65,20 €
    1 * 5 g
       65,20 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 301,00 €
    301,00 €
    1 * 25 g
       301,00 €