

2-Bromo-3'-(trifluoromethyl)acetophenone
Catalog # APOSPC5383-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:267.04 g/mol
Boiling Pt:64…72 °C/0.07mmHg
Melting Pt:20…23 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:2003-10-3
Formula:C₉H₆BrF₃O
MDL Number:MFCD03094283